Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014671.2 | NP_055486.2 | 18 - 19 | 2788 | 81 |
XM_005249564.4 | XP_005249621.1 | 17 - 18 | 2722 | 81 | |
XM_017012818.1 | XP_016868307.1 | 19 - 20 | 3060 | 81 | |
GenBank mRNA | AK127280.1 | - | 3 - 4 | 3059 | 81 |
AK295138.1 | - | 18 - 19 | 2468 | 81 | |
D13635.1 | - | 18 - 19 | 81 |