Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014671.2 | NP_055486.2 | 20 - 21 | 3188 | 68 |
XM_005249564.4 | XP_005249621.1 | 19 - 20 | 3122 | 68 | |
XM_017012818.1 | XP_016868307.1 | 21 - 22 | 3460 | 68 | |
GenBank mRNA | AK127280.1 | - | 5 - 6 | 3459 | 68 |
AK295138.1 | - | 20 - 21 | 2868 | 68 | |
D13635.1 | - | 20 - 21 | 3179 | 68 |