Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014671.2 | NP_055486.2 | 3 - 4 | 504 | 111 |
XM_017012818.1 | XP_016868307.1 | 4 - 5 | 776 | 111 | |
GenBank mRNA | AK295138.1 | - | 3 - 4 | 111 | |
BC014029.2 | - | 4 - 5 | 264 | 111 | |
D13635.1 | - | 3 - 4 | 495 | 111 |