Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014671.2 | NP_055486.2 | 8 - 9 | 1302 | 77 |
XM_005249564.4 | XP_005249621.1 | 7 - 8 | 1236 | 77 | |
XM_017012818.1 | XP_016868307.1 | 9 - 10 | 1574 | 77 | |
GenBank mRNA | AK295138.1 | - | 8 - 9 | 982 | 77 |
BC014029.2 | - | 9 - 10 | 1062 | 77 | |
BC026241.1 | - | 7 - 8 | 1177 | 77 | |
D13635.1 | - | 8 - 9 | 1293 | 77 |