Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253.3 | NP_001244.1 | 5 - 6 | 849 | 85 |
GenBank mRNA | AB007892.2 | - | 5 - 6 | 621 | 85 |
AK128737.1 | - | 5 - 6 | 657 | 85 | |
AK299738.1 | - | 5 - 6 | 576 | 85 | |
BC001568.1 | - | 5 - 6 | 638 | 85 | |
U86753.1 | - | 5 - 6 | 539 | 85 |