Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253.3 | NP_001244.1 | 7 - 8 | 1212 | 120 |
GenBank mRNA | AB007892.2 | - | 7 - 8 | 984 | 120 |
AK128737.1 | - | 7 - 8 | 1020 | 120 | |
AK299738.1 | - | 7 - 8 | 939 | 120 | |
BC001568.1 | - | 7 - 8 | 1001 | 120 | |
U86753.1 | - | 7 - 8 | 902 | 120 |