Assay Design



Gene Transcripts

Gene Symbol
WDR4
Entrez Gene ID
10785
Gene Name
WD repeat domain 4
Gene Aliases
TRM82, TRMT82
Chromosome Location
Chr.21: 42843075 - 42879865 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260476.1 NP_001247405.1 1 - 2 79 64
GenBank mRNA BC001074.2 - 1 - 2 58 64

View Full Product Details