Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007359.4 | NP_031385.2 | 10 - 11 | 2047 | 63 |
XM_005257163.1 | XP_005257220.1 | 10 - 11 | 1825 | 63 | |
GenBank mRNA | AK292425.1 | - | 10 - 11 | 1986 | 63 |
BC044656.1 | - | 10 - 11 | 1930 | 63 | |
BC050526.1 | - | 10 - 11 | 1950 | 63 | |
X80199.2 | - | 10 - 11 | 1985 | 63 |