Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040092.2 | NP_001035181.1 | 24 - 25 | 2511 | 84 |
NM_001130863.2 | NP_001124335.1 | 25 - 26 | 2586 | 84 | |
NM_006209.4 | NP_006200.3 | 25 - 26 | 2667 | 84 | |
NR_045555.1 | 13 - 14 | 1416 | 84 | ||
XM_006716584.1 | XP_006716647.1 | 26 - 27 | 2720 | 84 | |
XM_006716585.1 | XP_006716648.1 | 26 - 27 | 2708 | 84 | |
XM_006716587.1 | XP_006716650.1 | 25 - 26 | 2633 | 84 | |
XM_017013571.1 | XP_016869060.1 | 25 - 26 | 2620 | 84 | |
XM_017013572.1 | XP_016869061.1 | 25 - 26 | 2552 | 84 | |
XM_017013573.1 | XP_016869062.1 | 25 - 26 | 2608 | 84 | |
XM_017013574.1 | XP_016869063.1 | 24 - 25 | 2545 | 84 | |
XM_017013575.1 | XP_016869064.1 | 24 - 25 | 2477 | 84 | |
GenBank mRNA | AK124910.1 | - | 6 - 7 | 3046 | 84 |
AK130313.1 | - | 13 - 14 | 1416 | 84 | |
AK296027.1 | - | 10 - 11 | 1127 | 84 | |
AK315253.1 | - | 24 - 25 | 2484 | 84 | |
AL544867.3 | - | 7 - 8 | 813 | 84 | |
BC034961.2 | - | 24 - 25 | 2484 | 84 | |
D45421.1 | - | 24 - 25 | 2458 | 84 | |
EU131011.2 | - | 25 - 26 | 2500 | 84 | |
L35594.1 | - | 25 - 26 | 2630 | 84 |