Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015230.3 | NP_056045.2 | 3 - 4 | 1304 | 137 |
XM_017007698.1 | XP_016863187.1 | 3 - 4 | 1734 | 137 | |
XM_017007699.1 | XP_016863188.1 | 3 - 4 | 1189 | 137 | |
XM_017007700.1 | XP_016863189.1 | 3 - 4 | 1734 | 137 | |
XM_017007701.1 | XP_016863190.1 | 3 - 4 | 1734 | 137 | |
XM_017007702.1 | XP_016863191.1 | 3 - 4 | 1734 | 137 | |
GenBank mRNA | AB011152.2 | - | 2 - 3 | 747 | 137 |
AF439781.1 | - | 2 - 3 | 1126 | 137 | |
AK126864.1 | - | 3 - 4 | 1297 | 137 | |
AY049733.1 | - | 3 - 4 | 1219 | 137 | |
BC150258.1 | - | 2 - 3 | 758 | 137 |