Assay Design



Gene Transcripts

Gene Symbol
ARAP2
Entrez Gene ID
116984
Gene Name
ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 2
Gene Aliases
CENTD1, PARX
Chromosome Location
Chr.4: 36005186 - 36244832 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015230.3 NP_056045.2 11 - 12 2508 115
XM_017007698.1 XP_016863187.1 11 - 12 2938 115
XM_017007699.1 XP_016863188.1 11 - 12 2393 115
XM_017007700.1 XP_016863189.1 11 - 12 2938 115
XM_017007701.1 XP_016863190.1 11 - 12 2938 115
XM_017007702.1 XP_016863191.1 11 - 12 2938 115
GenBank mRNA AB011152.2 - 10 - 11 1951 115
AF439781.1 - 10 - 11 2330 115
AK126864.1 - 11 - 12 2501 115
AY049733.1 - 11 - 12 2423 115
BC150258.1 - 10 - 11 1962 115

View Full Product Details