Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278199.1 | NP_001265128.1 | 6 - 7 | 1305 | 100 |
NM_003100.3 | NP_003091.2 | 6 - 7 | 754 | 100 | |
GenBank mRNA | AF065482.1 | - | 6 - 7 | 675 | 100 |
AK002170.1 | - | 6 - 7 | 650 | 100 | |
AK023581.1 | - | 6 - 7 | 682 | 100 | |
AK023716.1 | - | 6 - 7 | 695 | 100 | |
AK293671.1 | - | 5 - 6 | 475 | 100 | |
AK296596.1 | - | 6 - 7 | 986 | 100 | |
AY272044.1 | - | 6 - 7 | 662 | 100 | |
BC003382.1 | - | 6 - 7 | 678 | 100 | |
BT009841.1 | - | 6 - 7 | 646 | 100 |