Assay Design



Gene Transcripts

Gene Symbol
SYNE2
Entrez Gene ID
23224
Gene Name
spectrin repeat containing nuclear envelope protein 2
Gene Aliases
EDMD5, NUA, NUANCE, Nesp2, Nesprin-2, SYNE-2, TROPH
Chromosome Location
Chr.14: 63761904 - 64226449 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015180.4 NP_055995.4 32 - 33 4957 104
NM_182914.2 NP_878918.2 32 - 33 4957 104
XM_005267454.1 XP_005267511.1 32 - 33 4957 104
XM_005267456.1 XP_005267513.1 32 - 33 4957 104
XM_005267457.1 XP_005267514.1 32 - 33 4957 104
XM_005267458.1 XP_005267515.1 32 - 33 4957 104
XM_005267459.1 XP_005267516.1 32 - 33 4957 104
XM_011536574.1 XP_011534876.1 32 - 33 4957 104
XM_011536575.2 XP_011534877.1 32 - 33 4934 104
XM_011536576.2 XP_011534878.1 33 - 34 5114 104
XM_011536577.2 XP_011534879.1 32 - 33 4881 104
XM_011536578.1 XP_011534880.1 32 - 33 4957 104
XM_011536579.1 XP_011534881.1 32 - 33 4957 104
XM_011536580.1 XP_011534882.1 32 - 33 4957 104
XM_011536581.1 XP_011534883.1 32 - 33 4957 104
XM_011536582.1 XP_011534884.1 32 - 33 4957 104
XM_011536584.2 XP_011534886.1 32 - 33 4957 104
XM_017021101.1 XP_016876590.1 32 - 33 4959 104
XM_017021102.1 XP_016876591.1 31 - 32 5002 104
GenBank mRNA AF357236.1 - 31 - 32 4727 104
AF435011.1 - 32 - 33 4939 104
AF495911.1 - 32 - 33 4924 104
AK125715.1 - 3 - 4 619 104
BX648234.1 - 3 - 4 637 104

View Full Product Details