Assay Design
Gene Transcripts
- Gene Symbol
- SYNE2
- Entrez Gene ID
-
23224
- Gene Name
- spectrin repeat containing nuclear envelope protein 2
- Gene Aliases
-
EDMD5, NUA, NUANCE, Nesp2, Nesprin-2, SYNE-2, TROPH
- Chromosome Location
-
Chr.14: 63761904 - 64226449 on Build GRCh38
View Full Product Details