Assay Design



Gene Transcripts

Gene Symbol
SYNE2
Entrez Gene ID
23224
Gene Name
spectrin repeat containing nuclear envelope protein 2
Gene Aliases
EDMD5, NUA, NUANCE, Nesp2, Nesprin-2, SYNE-2, TROPH
Chromosome Location
Chr.14: 63761904 - 64226449 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015180.4 NP_055995.4 96 - 97 17782 70
NM_182914.2 NP_878918.2 96 - 97 17782 70
XM_005267454.1 XP_005267511.1 96 - 97 17782 70
XM_005267456.1 XP_005267513.1 96 - 97 17782 70
XM_005267457.1 XP_005267514.1 96 - 97 17782 70
XM_005267458.1 XP_005267515.1 96 - 97 17782 70
XM_005267459.1 XP_005267516.1 96 - 97 17782 70
XM_011536574.1 XP_011534876.1 96 - 97 17782 70
XM_011536575.2 XP_011534877.1 96 - 97 17759 70
XM_011536576.2 XP_011534878.1 97 - 98 17939 70
XM_011536577.2 XP_011534879.1 96 - 97 17706 70
XM_011536578.1 XP_011534880.1 96 - 97 17782 70
XM_011536579.1 XP_011534881.1 96 - 97 17782 70
XM_011536580.1 XP_011534882.1 96 - 97 17782 70
XM_011536581.1 XP_011534883.1 96 - 97 17782 70
XM_011536582.1 XP_011534884.1 95 - 96 17665 70
XM_017021101.1 XP_016876590.1 96 - 97 17784 70
XM_017021102.1 XP_016876591.1 95 - 96 17827 70
GenBank mRNA AB023228.3 - 47 - 48 7454 70
AF435011.1 - 96 - 97 17764 70
AF495911.1 - 96 - 97 17749 70
AK074055.1 - 9 - 10 1416 70
AK090430.1 - 14 - 15 2158 70
AK095087.1 - 22 - 23 3520 70
AL080133.1 - 2 - 3 295 70
AY061759.1 - 44 - 45 7085 70
BX537642.1 - 17 - 18 2595 70
BX538095.1 - 5 - 6 716 70
CR749324.1 - 16 - 17 2479 70

View Full Product Details