Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135653.1 | NP_001129125.1 | 15 - 16 | 2898 | 112 |
NM_001135654.1 | NP_001129126.1 | 14 - 15 | 2811 | 112 | |
NM_003819.3 | NP_003810.1 | 15 - 16 | 2850 | 112 | |
GenBank mRNA | AB208962.1 | - | 9 - 10 | 1094 | 112 |
AK091110.1 | - | 3 - 4 | 1954 | 112 | |
AK222917.1 | - | 9 - 10 | 1135 | 112 | |
BC029969.1 | - | 15 - 16 | 2450 | 112 | |
BC065540.1 | - | 14 - 15 | 2714 | 112 | |
BC071591.1 | - | 15 - 16 | 2836 | 112 | |
BC094755.1 | - | 15 - 16 | 2839 | 112 | |
U33818.1 | - | 15 - 16 | 2105 | 112 | |
U75686.1 | - | 15 - 16 | 2746 | 112 |