Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135653.1 | NP_001129125.1 | 6 - 7 | 1772 | 153 |
NM_001135654.1 | NP_001129126.1 | 6 - 7 | 1772 | 153 | |
NM_003819.3 | NP_003810.1 | 6 - 7 | 1772 | 153 | |
GenBank mRNA | BC029969.1 | - | 6 - 7 | 1324 | 153 |
BC065540.1 | - | 6 - 7 | 1675 | 153 | |
BC071591.1 | - | 6 - 7 | 1710 | 153 | |
BC094755.1 | - | 6 - 7 | 1713 | 153 | |
BC106908.1 | - | 1 - 2 | 152 | 153 | |
BC118568.1 | - | 1 - 2 | 152 | 153 | |
D12799.1 | - | 3 - 4 | 307 | 153 | |
U33818.1 | - | 6 - 7 | 1027 | 153 | |
U75686.1 | - | 6 - 7 | 1668 | 153 |