Assay Design



Gene Transcripts

Gene Symbol
RCC1
Entrez Gene ID
1104
Gene Name
regulator of chromosome condensation 1
Gene Aliases
CHC1, RCC1-I, SNHG3-RCC1
Chromosome Location
Chr.1: 28505943 - 28539196 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001048194.2 NP_001041659.1 2 - 3 158 127
GenBank mRNA BC069198.1 - 2 - 3 123 127
KU177975.1 - 1 - 2 73 127

View Full Product Details