Assay Design



Gene Transcripts

Gene Symbol
HHLA1
Entrez Gene ID
10086
Gene Name
HERV-H LTR-associating 1
Gene Aliases
PLA2L
Chromosome Location
Chr.8: 132061486 - 132105265 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145095.1 NP_001138567.1 14 - 15 1461 123

View Full Product Details