Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205228.1 | NP_001192157.1 | 3 - 4 | 460 | 151 |
NM_002959.6 | NP_002950.3 | 3 - 4 | 488 | 151 | |
XM_005271100.2 | XP_005271157.1 | 3 - 4 | 483 | 151 | |
XM_005271101.2 | XP_005271158.1 | 3 - 4 | 602 | 151 | |
XM_005271102.1 | XP_005271159.1 | 3 - 4 | 522 | 151 | |
XM_006710812.1 | XP_006710875.1 | 3 - 4 | 258 | 151 | |
GenBank mRNA | AK293118.1 | - | 3 - 4 | 446 | 151 |
AK301548.1 | - | 3 - 4 | 463 | 151 | |
AK310147.1 | - | 3 - 4 | 351 | 151 | |
BC023542.1 | - | 3 - 4 | 484 | 151 | |
DB045034.1 | - | 3 - 4 | 200 | 151 |