Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000423.2 | NP_000414.2 | 5 - 6 | 1156 | 76 |
GenBank mRNA | BC096294.4 | - | 5 - 6 | 1145 | 76 |
BC099643.1 | - | 5 - 6 | 1145 | 76 | |
BC099644.3 | - | 5 - 6 | 1145 | 76 | |
M99061.1 | - | 5 - 6 | 1174 | 76 |