Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288718.1 | NP_001275647.1 | 18 - 19 | 2962 | 117 |
NM_001288719.1 | NP_001275648.1 | 17 - 18 | 2683 | 117 | |
NM_001288720.1 | NP_001275649.1 | 17 - 18 | 2680 | 117 | |
NM_003152.3 | NP_003143.2 | 19 - 20 | 2862 | 117 | |
GenBank mRNA | AB208972.1 | - | 17 - 18 | 2193 | 117 |
AK291650.1 | - | 18 - 19 | 2962 | 117 | |
AK301457.1 | - | 17 - 18 | 2241 | 117 | |
BC027036.1 | - | 18 - 19 | 2330 | 117 | |
DQ471288.1 | - | 17 - 18 | 2661 | 117 | |
L41142.1 | - | 18 - 19 | 2277 | 117 | |
U43185.1 | - | 19 - 20 | 2860 | 117 |