Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001621.4 | NP_001612.1 | 10 - 11 | 3019 | 106 |
GenBank mRNA | BC069390.1 | - | 10 - 11 | 2476 | 106 |
BC070080.1 | - | 10 - 11 | 2937 | 106 | |
D16354.1 | - | 10 - 11 | 2988 | 107 | |
L19872.1 | - | 10 - 11 | 2781 | 106 |