Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000410.3 | NP_000401.1 | 4 - 5 | 1053 | 74 |
NM_001300749.1 | NP_001287678.1 | 4 - 5 | 1053 | 74 | |
NM_139003.2 | NP_620572.1 | 3 - 4 | 735 | 74 | |
NM_139004.2 | NP_620573.1 | 3 - 4 | 777 | 74 | |
NM_139006.2 | NP_620575.1 | 4 - 5 | 1011 | 74 | |
NM_139007.2 | NP_620576.1 | 3 - 4 | 789 | 74 | |
NM_139008.2 | NP_620577.1 | 3 - 4 | 747 | 74 | |
NM_139009.2 | NP_620578.1 | 4 - 5 | 984 | 74 | |
NM_139010.2 | NP_620579.1 | 2 - 3 | 513 | 74 | |
XM_011514543.2 | XP_011512845.1 | 4 - 5 | 1015 | 74 | |
GenBank mRNA | AF079407.1 | - | 4 - 5 | 887 | 74 |
AF079408.1 | - | 3 - 4 | 665 | 74 | |
AF079409.1 | - | 3 - 4 | 623 | 74 | |
AF115265.1 | - | 4 - 5 | 904 | 74 | |
AJ249335.1 | - | 4 - 5 | 984 | 74 | |
AJ249336.1 | - | 3 - 4 | 789 | 74 | |
AJ249337.1 | - | 3 - 4 | 777 | 74 | |
AJ250635.1 | - | 2 - 3 | 513 | 74 | |
AK300933.1 | - | 4 - 5 | 897 | 74 | |
AY205604.1 | - | 3 - 4 | 654 | 74 | |
BC074721.2 | - | 5 - 6 | 933 | 74 | |
BC117201.1 | - | 4 - 5 | 928 | 74 | |
BC117203.1 | - | 4 - 5 | 928 | 74 | |
JN375755.1 | - | 1 - 2 | 111 | 74 | |
JN375756.1 | - | 1 - 2 | 110 | 74 | |
JN375757.1 | - | 1 - 2 | 101 | 74 | |
JN375758.1 | - | 1 - 2 | 110 | 74 | |
JN375759.1 | - | 1 - 2 | 116 | 74 | |
U60319.1 | - | 4 - 5 | 1114 | 74 |