Assay Design



Gene Transcripts

Gene Symbol
DNAJB5
Entrez Gene ID
25822
Gene Name
DnaJ heat shock protein family (Hsp40) member B5
Gene Aliases
Hsc40
Chromosome Location
Chr.9: 34989369 - 34998433 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012266.5 NP_036398.3 1 - 2 189 122
GenBank mRNA AK023253.1 - 1 - 2 85 122
AK314671.1 - 1 - 2 66 122
BU845478.1 - 1 - 2 102 122

View Full Product Details