Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004593.2 | NP_004584.1 | 2 - 3 | 442 | 69 |
XM_005247703.1 | XP_005247760.1 | 3 - 4 | 610 | 69 | |
XM_011513072.2 | XP_011511374.1 | 3 - 4 | 3412 | 69 | |
XM_017007033.1 | XP_016862522.1 | 2 - 3 | 1588 | 69 | |
GenBank mRNA | AB209208.1 | - | 3 - 4 | 3399 | 69 |
AK315632.1 | - | 2 - 3 | 426 | 69 | |
BC000160.2 | - | 2 - 3 | 309 | 69 | |
BC000451.2 | - | 2 - 3 | 308 | 69 | |
BC005898.1 | - | 2 - 3 | 293 | 69 | |
CR749214.1 | - | 1 - 2 | 2469 | 69 | |
U61267.1 | - | 2 - 3 | 287 | 69 | |
U68063.1 | - | 2 - 3 | 295 | 69 |