Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243879.1 | NP_001230808.1 | 4 - 5 | 785 | 85 |
NM_004593.2 | NP_004584.1 | 5 - 6 | 919 | 85 | |
XM_005247703.1 | XP_005247760.1 | 6 - 7 | 1087 | 85 | |
XM_011513072.2 | XP_011511374.1 | 6 - 7 | 3889 | 85 | |
XM_017007033.1 | XP_016862522.1 | 5 - 6 | 2065 | 85 | |
GenBank mRNA | AB209208.1 | - | 6 - 7 | 3876 | 85 |
AK091863.1 | - | 1 - 2 | 1806 | 85 | |
AK094495.1 | - | 2 - 3 | 841 | 85 | |
AK098191.1 | - | 5 - 6 | 706 | 85 | |
AK125610.1 | - | 2 - 3 | 1589 | 85 | |
AK301118.1 | - | 4 - 5 | 638 | 85 | |
AK308267.1 | - | 4 - 5 | 663 | 85 | |
AK315632.1 | - | 5 - 6 | 903 | 85 | |
BC000160.2 | - | 5 - 6 | 786 | 85 | |
BC000451.2 | - | 5 - 6 | 785 | 85 | |
BC005898.1 | - | 5 - 6 | 770 | 85 | |
CR749214.1 | - | 4 - 5 | 2946 | 85 | |
U61267.1 | - | 5 - 6 | 764 | 85 | |
U68063.1 | - | 5 - 6 | 772 | 85 |