Assay Design



Gene Transcripts

Gene Symbol
PRPF4B
Entrez Gene ID
8899
Gene Name
pre-mRNA processing factor 4B
Gene Aliases
PR4H, PRP4, PRP4H, PRP4K, dJ1013A10.1
Chromosome Location
Chr.6: 4019402 - 4064983 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003913.4 NP_003904.3 13 - 14 2829 145
XM_017011410.1 XP_016866899.1 13 - 14 4762 145
XM_017011411.1 XP_016866900.1 13 - 14 4762 145
XM_017011412.1 XP_016866901.1 13 - 14 4762 145
XM_017011413.1 XP_016866902.1 13 - 14 4762 145
XM_017011414.1 XP_016866903.1 13 - 14 4762 145
XM_017011415.1 XP_016866904.1 13 - 14 4762 145
XM_017011416.1 XP_016866905.1 13 - 14 4762 145
XM_017011417.1 XP_016866906.1 13 - 14 4762 145
XM_017011418.1 XP_016866907.1 13 - 14 4762 145
XM_017011419.1 XP_016866908.1 13 - 14 4762 145
XM_017011420.1 XP_016866909.1 13 - 14 4762 145
XM_017011421.1 XP_016866910.1 13 - 14 4762 145
XM_017011422.1 XP_016866911.1 13 - 14 4762 145
XM_017011423.1 XP_016866912.1 13 - 14 4762 145
XM_017011424.1 XP_016866913.1 13 - 14 4762 145
XM_017011425.1 XP_016866914.1 13 - 14 4762 145
XM_017011426.1 XP_016866915.1 13 - 14 4762 145
XM_017011427.1 XP_016866916.1 13 - 14 4762 145
XM_017011428.1 XP_016866917.1 13 - 14 4762 145
XM_017011429.1 XP_016866918.1 13 - 14 4762 145
XM_017011430.1 XP_016866919.1 13 - 14 4762 145
XM_017011431.1 XP_016866920.1 13 - 14 4762 145
XM_017011432.1 XP_016866921.1 14 - 15 4924 145
XM_017011433.1 XP_016866922.1 14 - 15 4924 145
XM_017011434.1 XP_016866923.1 14 - 15 4924 145
XM_017011435.1 XP_016866924.1 14 - 15 4924 145
GenBank mRNA AB011108.1 - 13 - 14 2802 145
AF283465.1 - 13 - 14 2780 145
AK291244.1 - 13 - 14 2807 145
AK303047.1 - 13 - 14 2854 145
AY029347.1 - 13 - 14 2738 145
BC009844.2 - 11 - 12 1477 145
BC034969.1 - 13 - 14 2842 145
U48736.1 - 10 - 11 1318 145

View Full Product Details