Assay Design



Gene Transcripts

Gene Symbol
PRPF4B
Entrez Gene ID
8899
Gene Name
pre-mRNA processing factor 4B
Gene Aliases
PR4H, PRP4, PRP4H, PRP4K, dJ1013A10.1
Chromosome Location
Chr.6: 4019402 - 4064983 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003913.4 NP_003904.3 6 - 7 1952 139
XM_011514970.2 XP_011513272.1 6 - 7 3885 139
XM_017011410.1 XP_016866899.1 6 - 7 3885 139
XM_017011411.1 XP_016866900.1 6 - 7 3885 139
XM_017011412.1 XP_016866901.1 6 - 7 3885 139
XM_017011413.1 XP_016866902.1 6 - 7 3885 139
XM_017011414.1 XP_016866903.1 6 - 7 3885 139
XM_017011415.1 XP_016866904.1 6 - 7 3885 139
XM_017011416.1 XP_016866905.1 6 - 7 3885 139
XM_017011417.1 XP_016866906.1 6 - 7 3885 139
XM_017011418.1 XP_016866907.1 6 - 7 3885 139
XM_017011419.1 XP_016866908.1 6 - 7 3885 139
XM_017011420.1 XP_016866909.1 6 - 7 3885 139
XM_017011421.1 XP_016866910.1 6 - 7 3885 139
XM_017011422.1 XP_016866911.1 6 - 7 3885 139
XM_017011423.1 XP_016866912.1 6 - 7 3885 139
XM_017011424.1 XP_016866913.1 6 - 7 3885 139
XM_017011425.1 XP_016866914.1 6 - 7 3885 139
XM_017011426.1 XP_016866915.1 6 - 7 3885 139
XM_017011427.1 XP_016866916.1 6 - 7 3885 139
XM_017011428.1 XP_016866917.1 6 - 7 3885 139
XM_017011429.1 XP_016866918.1 6 - 7 3885 139
XM_017011430.1 XP_016866919.1 6 - 7 3885 139
XM_017011431.1 XP_016866920.1 6 - 7 3885 139
XM_017011432.1 XP_016866921.1 6 - 7 3885 139
XM_017011433.1 XP_016866922.1 6 - 7 3885 139
XM_017011434.1 XP_016866923.1 6 - 7 3885 139
XM_017011435.1 XP_016866924.1 6 - 7 3885 139
GenBank mRNA AB011108.1 - 6 - 7 1925 139
AF283465.1 - 6 - 7 1903 139
AK291244.1 - 6 - 7 1930 139
AK303047.1 - 6 - 7 1977 139
AY029347.1 - 6 - 7 1861 139
BC009844.2 - 4 - 5 600 139
BC034969.1 - 6 - 7 1965 139
U48736.1 - 3 - 4 441 139

View Full Product Details