Assay Design



Gene Transcripts

Gene Symbol
CRTAC1
Entrez Gene ID
55118
Gene Name
cartilage acidic protein 1
Gene Aliases
ASPIC, ASPIC1, CEP-68
Chromosome Location
Chr.10: 97854711 - 98030828 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206528.2 NP_001193457.1 5 - 6 1075 141
NM_018058.6 NP_060528.3 5 - 6 1075 141
XM_005269938.4 XP_005269995.1 5 - 6 868 141
XM_011539917.1 XP_011538219.1 5 - 6 868 141
XM_017016366.1 XP_016871855.1 5 - 6 868 141
XM_017016367.1 XP_016871856.1 5 - 6 868 141
GenBank mRNA AJ276171.1 - 5 - 6 868 141
AJ279016.1 - 4 - 5 695 141
AJ421515.1 - 4 - 5 695 141
AK001182.1 - 3 - 4 407 141
AK057190.1 - 5 - 6 1075 141
AK292084.1 - 5 - 6 837 141
BC034245.1 - 3 - 4 337 141

View Full Product Details