Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004615.3 | NP_004606.2 | 2 - 3 | 341 | 59 |
GenBank mRNA | AB062057.1 | - | 2 - 3 | 342 | 59 |
AB102665.1 | - | 2 - 3 | 257 | 59 | |
AK293178.1 | - | 3 - 4 | 326 | 59 | |
AK293534.1 | - | 4 - 5 | 495 | 59 | |
AK293552.1 | - | 3 - 4 | 406 | 59 | |
AK312343.1 | - | 2 - 3 | 275 | 59 | |
BC018036.1 | - | 2 - 3 | 300 | 59 | |
D10653.1 | - | 2 - 3 | 281 | 59 | |
D29808.1 | - | 2 - 3 | 267 | 59 | |
L10373.1 | - | 2 - 3 | 325 | 59 |