Assay Design



Gene Transcripts

Gene Symbol
STX5
Entrez Gene ID
6811
Gene Name
syntaxin 5
Gene Aliases
SED5, STX5A
Chromosome Location
Chr.11: 62806860 - 62832091 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244666.1 NP_001231595.1 3 - 4 453 77
NM_003164.4 NP_003155.2 3 - 4 453 77
XM_011545223.2 XP_011543525.1 3 - 4 278 77
XM_011545224.2 XP_011543526.1 2 - 3 169 77
GenBank mRNA AK296675.1 - 2 - 3 169 77
AK313497.1 - 3 - 4 413 77
BC002645.1 - 3 - 4 403 77
BC012137.1 - 3 - 4 430 77
BT019646.1 - 2 - 3 134 77
BT019647.1 - 2 - 3 134 77
BX537426.1 - 3 - 4 451 77
CB155395.1 - 3 - 4 453 77
U26648.1 - 2 - 3 160 77

View Full Product Details