Assay Design



Gene Transcripts

Gene Symbol
STX5
Entrez Gene ID
6811
Gene Name
syntaxin 5
Gene Aliases
SED5, STX5A
Chromosome Location
Chr.11: 62806860 - 62832091 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244666.1 NP_001231595.1 7 - 8 756 98
NM_003164.4 NP_003155.2 7 - 8 756 98
XM_011545223.2 XP_011543525.1 7 - 8 581 98
XM_011545224.2 XP_011543526.1 6 - 7 472 98
GenBank mRNA AK296675.1 - 6 - 7 472 98
AK313497.1 - 7 - 8 716 98
BC002645.1 - 7 - 8 706 98
BC012137.1 - 7 - 8 733 98
BT019646.1 - 6 - 7 437 98
BT019647.1 - 6 - 7 437 98
BX537426.1 - 7 - 8 754 98
U26648.1 - 6 - 7 463 98

View Full Product Details