Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003164.4 | NP_003155.2 | 10 - 11 | 1068 | 65 |
XM_011545223.2 | XP_011543525.1 | 10 - 11 | 893 | 65 | |
XM_011545224.2 | XP_011543526.1 | 9 - 10 | 784 | 65 | |
GenBank mRNA | AK296675.1 | - | 9 - 10 | 784 | 65 |
AK313497.1 | - | 10 - 11 | 1028 | 65 | |
BC012137.1 | - | 10 - 11 | 1045 | 65 | |
BT019646.1 | - | 9 - 10 | 749 | 65 | |
BT019647.1 | - | 9 - 10 | 749 | 65 | |
BX537426.1 | - | 10 - 11 | 1066 | 65 | |
U26648.1 | - | 9 - 10 | 775 | 65 |