Assay Design



Gene Transcripts

Gene Symbol
NEIL1
Entrez Gene ID
79661
Gene Name
nei like DNA glycosylase 1
Gene Aliases
FPG1, NEI1, hFPG1
Chromosome Location
Chr.15: 75346895 - 75355251 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256552.1 NP_001243481.1 9 - 10 1532 91
NM_024608.3 NP_078884.2 9 - 10 1608 91
NR_046311.1 10 - 11 1731 91
XM_005254659.4 XP_005254716.1 9 - 10 1468 91
XM_006720677.3 XP_006720740.1 9 - 10 1453 91
XM_006720678.3 XP_006720741.1 8 - 9 1363 91
XM_006720679.3 XP_006720742.1 9 - 10 1391 91
XM_006720680.1 XP_006720743.1 10 - 11 1623 91
XM_006720681.1 XP_006720744.1 10 - 11 1470 91
XM_011522001.2 XP_011520303.2 9 - 10 1373 91
XM_011522002.1 XP_011520304.1 10 - 11 1596 91
XM_011522003.2 XP_011520305.1 11 - 12 1652 91
XM_011522004.2 XP_011520306.1 9 - 10 1340 91
XM_011522005.1 XP_011520307.1 10 - 11 1682 91
XM_011522006.1 XP_011520308.1 9 - 10 1606 91
XM_011522007.1 XP_011520309.1 10 - 11 1702 91
XM_011522008.1 XP_011520310.1 10 - 11 1675 91
XM_017022566.1 XP_016878055.1 8 - 9 1335 91
XM_017022567.1 XP_016878056.1 9 - 10 1366 91
GenBank mRNA AB079068.1 - 8 - 9 1102 91
AK026055.1 - 10 - 11 1529 91
AK026216.1 - 3 - 4 885 91
AK097008.1 - 10 - 11 1731 91
AK128372.1 - 7 - 8 2776 91
BC010876.1 - 9 - 10 1513 91

View Full Product Details