Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039702.2 | NP_001034791.1 | 5 - 6 | 663 | 140 |
NM_018324.2 | NP_060794.1 | 6 - 7 | 822 | 140 | |
XM_017016376.1 | XP_016871865.1 | 6 - 7 | 786 | 140 | |
GenBank mRNA | AK001844.1 | - | 5 - 6 | 613 | 140 |
AK001968.1 | - | 6 - 7 | 755 | 140 | |
BC050372.1 | - | 5 - 6 | 630 | 140 |