Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145944.1 | NP_001139416.1 | 9 - 10 | 1622 | 108 |
NM_001145945.1 | NP_001139417.1 | 11 - 12 | 1758 | 108 | |
NM_001145946.1 | NP_001139418.1 | 12 - 13 | 1905 | 108 | |
NM_017435.4 | NP_059131.1 | 11 - 12 | 1918 | 108 | |
XM_005253394.2 | XP_005253451.1 | 12 - 13 | 1694 | 108 | |
XM_005253396.2 | XP_005253453.1 | 10 - 11 | 6459 | 108 | |
XM_011520703.2 | XP_011519005.1 | 12 - 13 | 1694 | 108 | |
XM_011520704.2 | XP_011519006.1 | 12 - 13 | 1694 | 108 | |
XM_011520709.2 | XP_011519011.1 | 9 - 10 | 1336 | 108 | |
XM_011520711.2 | XP_011519013.1 | 11 - 12 | 6514 | 108 | |
XM_017019484.1 | XP_016874973.1 | 14 - 15 | 6955 | 108 | |
XM_017019485.1 | XP_016874974.1 | 13 - 14 | 6873 | 108 | |
XM_017019486.1 | XP_016874975.1 | 11 - 12 | 6613 | 108 | |
XM_017019487.1 | XP_016874976.1 | 10 - 11 | 6459 | 108 | |
XM_017019488.1 | XP_016874977.1 | 15 - 16 | 7010 | 108 | |
XM_017019489.1 | XP_016874978.1 | 16 - 17 | 7110 | 108 | |
XM_017019490.1 | XP_016874979.1 | 16 - 17 | 7074 | 108 | |
GenBank mRNA | AF205076.1 | - | 11 - 12 | 1919 | 108 |
AF260704.1 | - | 11 - 12 | 1614 | 108 | |
AK294333.1 | - | 9 - 10 | 1490 | 108 | |
AK296236.1 | - | 11 - 12 | 1651 | 108 | |
AK303713.1 | - | 12 - 13 | 1781 | 108 | |
AK308729.1 | - | 10 - 11 | 1554 | 108 | |
BC022461.2 | - | 11 - 12 | 1647 | 108 |