Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145945.1 | NP_001139417.1 | 4 - 5 | 629 | 76 |
NM_001145946.1 | NP_001139418.1 | 4 - 5 | 629 | 76 | |
NM_017435.4 | NP_059131.1 | 3 - 4 | 642 | 76 | |
XM_005253394.2 | XP_005253451.1 | 4 - 5 | 418 | 76 | |
XM_011520703.2 | XP_011519005.1 | 4 - 5 | 418 | 76 | |
XM_011520704.2 | XP_011519006.1 | 4 - 5 | 418 | 76 | |
GenBank mRNA | AF205076.1 | - | 3 - 4 | 643 | 76 |
AF260704.1 | - | 3 - 4 | 338 | 76 | |
AK296236.1 | - | 4 - 5 | 522 | 76 | |
AK303713.1 | - | 4 - 5 | 505 | 76 | |
AL834209.1 | - | 4 - 5 | 494 | 76 | |
BC022461.2 | - | 3 - 4 | 371 | 76 |