Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145944.1 | NP_001139416.1 | 4 - 5 | 751 | 85 |
NM_001145946.1 | NP_001139418.1 | 7 - 8 | 1034 | 85 | |
NM_017435.4 | NP_059131.1 | 6 - 7 | 1047 | 85 | |
XM_005253394.2 | XP_005253451.1 | 7 - 8 | 823 | 85 | |
XM_005253396.2 | XP_005253453.1 | 5 - 6 | 5588 | 85 | |
XM_011520703.2 | XP_011519005.1 | 7 - 8 | 823 | 85 | |
XM_011520704.2 | XP_011519006.1 | 7 - 8 | 823 | 85 | |
XM_011520709.2 | XP_011519011.1 | 4 - 5 | 465 | 85 | |
XM_011520711.2 | XP_011519013.1 | 6 - 7 | 5643 | 85 | |
XM_017019484.1 | XP_016874973.1 | 9 - 10 | 6084 | 85 | |
XM_017019485.1 | XP_016874974.1 | 8 - 9 | 6002 | 85 | |
XM_017019486.1 | XP_016874975.1 | 6 - 7 | 5742 | 85 | |
XM_017019487.1 | XP_016874976.1 | 5 - 6 | 5588 | 85 | |
XM_017019488.1 | XP_016874977.1 | 10 - 11 | 6139 | 85 | |
XM_017019489.1 | XP_016874978.1 | 11 - 12 | 6239 | 85 | |
XM_017019490.1 | XP_016874979.1 | 11 - 12 | 6203 | 85 | |
GenBank mRNA | AF205076.1 | - | 6 - 7 | 1048 | 85 |
AF260704.1 | - | 6 - 7 | 743 | 85 | |
AK294333.1 | - | 4 - 5 | 619 | 85 | |
AK303713.1 | - | 7 - 8 | 910 | 85 | |
AK308729.1 | - | 5 - 6 | 683 | 85 | |
AL834209.1 | - | 7 - 8 | 899 | 85 | |
BC022461.2 | - | 6 - 7 | 776 | 85 |