Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009813.2 | NP_001009813.1 | 11 - 12 | 1476 | 126 |
NM_001301059.1 | NP_001287988.1 | 11 - 12 | 1527 | 126 | |
NM_020160.2 | NP_064545.1 | 11 - 12 | 1665 | 126 | |
XM_011527136.2 | XP_011525438.1 | 11 - 12 | 1674 | 126 | |
XM_011527138.2 | XP_011525440.1 | 9 - 10 | 1524 | 126 | |
XM_011527139.2 | XP_011525441.1 | 11 - 12 | 1195 | 126 | |
XM_011527140.1 | XP_011525442.1 | 11 - 12 | 1694 | 126 | |
XM_017027011.1 | XP_016882500.1 | 11 - 12 | 1725 | 126 | |
GenBank mRNA | AB209455.1 | - | 6 - 7 | 705 | 126 |
AK098710.1 | - | 11 - 12 | 1389 | 126 | |
AK289950.1 | - | 11 - 12 | 1259 | 126 | |
BC025404.1 | - | 11 - 12 | 1428 | 126 | |
BC045741.1 | - | 4 - 5 | 617 | 126 | |
BC069251.1 | - | 11 - 12 | 1098 | 126 | |
BI522513.1 | - | 4 - 5 | 320 | 126 |