Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009813.2 | NP_001009813.1 | 6 - 7 | 993 | 79 |
NM_001301059.1 | NP_001287988.1 | 6 - 7 | 1044 | 79 | |
NM_020160.2 | NP_064545.1 | 6 - 7 | 1044 | 79 | |
XM_011527136.2 | XP_011525438.1 | 6 - 7 | 1191 | 79 | |
XM_011527139.2 | XP_011525441.1 | 6 - 7 | 712 | 79 | |
XM_011527140.1 | XP_011525442.1 | 6 - 7 | 1211 | 79 | |
XM_017027011.1 | XP_016882500.1 | 6 - 7 | 1242 | 79 | |
GenBank mRNA | AB209455.1 | - | 1 - 2 | 222 | 79 |
AK098710.1 | - | 6 - 7 | 906 | 79 | |
AK289950.1 | - | 6 - 7 | 638 | 79 | |
AL537583.3 | - | 6 - 7 | 785 | 79 | |
AY091593.1 | - | 4 - 5 | 273 | 79 | |
BC025404.1 | - | 6 - 7 | 807 | 79 | |
BC069251.1 | - | 6 - 7 | 615 | 79 | |
BM783223.1 | - | 4 - 5 | 420 | 79 |