Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003884.4 | NP_003875.3 | 12 - 13 | 2311 | 138 |
XM_005265528.4 | XP_005265585.1 | 12 - 13 | 1936 | 138 | |
XM_017007423.1 | XP_016862912.1 | 13 - 14 | 1766 | 138 | |
XM_017007424.1 | XP_016862913.1 | 12 - 13 | 1862 | 138 | |
GenBank mRNA | AL832173.1 | - | 2 - 3 | 224 | 138 |
BC060823.1 | - | 12 - 13 | 1916 | 138 | |
BC070075.1 | - | 12 - 13 | 1981 | 138 | |
U57317.2 | - | 12 - 13 | 2323 | 138 |