Assay Design



Gene Transcripts

Gene Symbol
MBTD1
Entrez Gene ID
54799
Gene Name
mbt domain containing 1
Gene Aliases
SA49P01
Chromosome Location
Chr.17: 51177419 - 51260511 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011524933.2 XP_011523235.1 6 - 7 832 71
GenBank mRNA AK000062.1 - 1 - 2 42 71

View Full Product Details