Assay Design



Gene Transcripts

Gene Symbol
SMURF2
Entrez Gene ID
64750
Gene Name
SMAD specific E3 ubiquitin protein ligase 2
Gene Aliases
-
Chromosome Location
Chr.17: 64542282 - 64662268 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022739.3 NP_073576.1 18 - 19 2537 112
XM_005257585.3 XP_005257642.1 17 - 18 2137 112
XM_011525128.1 XP_011523430.1 18 - 19 2155 112
GenBank mRNA AF301463.1 - 18 - 19 2157 112
AF310676.1 - 18 - 19 2149 112
AY014180.1 - 18 - 19 2337 112
BC009527.1 - 6 - 7 771 112
BC093876.1 - 18 - 19 2219 112
BC111945.1 - 18 - 19 2221 112
CR936721.1 - 19 - 20 2272 112

View Full Product Details