Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138424.1 | NP_612433.1 | 10 - 11 | 1134 | 70 |
XM_005251683.4 | XP_005251740.1 | 13 - 14 | 2123 | 70 | |
XM_006716947.2 | XP_006717010.1 | 12 - 13 | 1375 | 70 | |
XM_011518190.2 | XP_011516492.1 | 13 - 14 | 2158 | 70 | |
XM_011518191.2 | XP_011516493.1 | 13 - 14 | 2157 | 70 | |
XM_011518192.2 | XP_011516494.1 | 13 - 14 | 2155 | 70 | |
XM_017014240.1 | XP_016869729.1 | 7 - 8 | 814 | 70 | |
GenBank mRNA | BC010626.1 | - | 10 - 11 | 1134 | 70 |