Assay Design



Gene Transcripts

Gene Symbol
KIF12
Entrez Gene ID
113220
Gene Name
kinesin family member 12
Gene Aliases
-
Chromosome Location
Chr.9: 114091623 - 114100099 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138424.1 NP_612433.1 12 - 13 1314 93
XM_005251683.4 XP_005251740.1 15 - 16 2303 93
XM_006716947.2 XP_006717010.1 14 - 15 1555 93
XM_011518190.2 XP_011516492.1 15 - 16 2338 93
XM_011518191.2 XP_011516493.1 15 - 16 2337 93
XM_011518192.2 XP_011516494.1 15 - 16 2335 93
XM_017014240.1 XP_016869729.1 9 - 10 994 93
GenBank mRNA BC010626.1 - 12 - 13 1314 93

View Full Product Details