Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145945.1 | NP_001139417.1 | 1 - 2 | 252 | 65 |
NM_001145946.1 | NP_001139418.1 | 1 - 2 | 252 | 65 | |
XM_005253394.2 | XP_005253451.1 | 1 - 2 | 41 | 65 | |
XM_005253396.2 | XP_005253453.1 | 1 - 2 | 5102 | 65 | |
XM_011520703.2 | XP_011519005.1 | 1 - 2 | 41 | 65 | |
XM_011520704.2 | XP_011519006.1 | 1 - 2 | 41 | 65 | |
XM_011520711.2 | XP_011519013.1 | 1 - 2 | 5102 | 65 | |
XM_017019484.1 | XP_016874973.1 | 1 - 2 | 5102 | 65 | |
XM_017019485.1 | XP_016874974.1 | 1 - 2 | 5102 | 65 | |
XM_017019486.1 | XP_016874975.1 | 1 - 2 | 5102 | 65 | |
XM_017019487.1 | XP_016874976.1 | 1 - 2 | 5102 | 65 | |
XM_017019488.1 | XP_016874977.1 | 1 - 2 | 5102 | 65 | |
XM_017019489.1 | XP_016874978.1 | 1 - 2 | 5102 | 65 | |
XM_017019490.1 | XP_016874979.1 | 1 - 2 | 5102 | 65 | |
GenBank mRNA | AK296236.1 | - | 1 - 2 | 145 | 65 |
AK303713.1 | - | 1 - 2 | 128 | 65 | |
AK308729.1 | - | 1 - 2 | 176 | 65 | |
AL834209.1 | - | 1 - 2 | 117 | 65 | |
DA345251.1 | - | 1 - 2 | 252 | 65 |