Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003826.2 | NP_003817.1 | 8 - 9 | 733 | 82 |
XM_011525754.2 | XP_011524056.1 | 9 - 10 | 817 | 82 | |
XM_011525756.2 | XP_011524058.1 | 6 - 7 | 372 | 82 | |
XM_017026063.1 | XP_016881552.1 | 4 - 5 | 289 | 82 | |
GenBank mRNA | AK126942.1 | - | 3 - 4 | 886 | 82 |
AK294038.1 | - | 7 - 8 | 512 | 82 | |
AK314495.1 | - | 8 - 9 | 572 | 82 | |
BC001889.1 | - | 8 - 9 | 569 | 82 | |
CR536554.1 | - | 8 - 9 | 506 | 82 | |
U78107.1 | - | 8 - 9 | 579 | 82 |