Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098477.1 | NP_001091947.1 | 5 - 6 | 865 | 79 |
NM_002092.3 | NP_002083.3 | 5 - 6 | 1029 | 79 | |
XM_005265681.3 | XP_005265738.1 | 5 - 6 | 1268 | 79 | |
XM_011531897.2 | XP_011530199.1 | 5 - 6 | 694 | 79 | |
GenBank mRNA | AK055499.1 | - | 6 - 7 | 805 | 79 |
AK056891.1 | - | 6 - 7 | 775 | 79 | |
AK090755.1 | - | 6 - 7 | 809 | 79 | |
AK094806.1 | - | 5 - 6 | 834 | 79 | |
AK095799.1 | - | 6 - 7 | 795 | 79 | |
AK097055.1 | - | 6 - 7 | 875 | 79 | |
AK124124.1 | - | 6 - 7 | 946 | 79 | |
AK127161.1 | - | 6 - 7 | 904 | 79 | |
AK294612.1 | - | 5 - 6 | 729 | 79 | |
AK296287.1 | - | 6 - 7 | 864 | 79 | |
AK298883.1 | - | 5 - 6 | 687 | 79 | |
U07231.1 | - | 6 - 7 | 839 | 79 |