Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199173.1 | NP_001186102.1 | 8 - 9 | 1221 | 81 |
NM_001199174.1 | NP_001186103.1 | 8 - 9 | 1060 | 81 | |
NM_001199175.1 | NP_001186104.1 | 8 - 9 | 1248 | 81 | |
NM_022372.4 | NP_071767.3 | 8 - 9 | 1251 | 81 | |
XM_005255475.3 | XP_005255532.2 | 8 - 9 | 1027 | 81 | |
XM_005255478.2 | XP_005255535.1 | 8 - 9 | 1232 | 81 | |
XM_005255479.2 | XP_005255536.1 | 8 - 9 | 1159 | 81 | |
XM_017023548.1 | XP_016879037.1 | 8 - 9 | 1216 | 81 | |
XM_017023549.1 | XP_016879038.1 | 8 - 9 | 957 | 81 | |
XM_017023551.1 | XP_016879040.1 | 8 - 9 | 1266 | 81 | |
GenBank mRNA | AF195883.1 | - | 5 - 6 | 572 | 81 |
AK021536.1 | - | 8 - 9 | 1220 | 81 | |
AK022227.1 | - | 7 - 8 | 905 | 81 | |
AK098762.1 | - | 8 - 9 | 1156 | 81 | |
AK123195.1 | - | 1 - 2 | 1026 | 81 | |
AK302201.1 | - | 7 - 8 | 1060 | 81 | |
AY223837.1 | - | 7 - 8 | 1061 | 81 | |
BC001313.2 | - | 8 - 9 | 941 | 81 | |
BC017119.1 | - | 7 - 8 | 952 | 81 | |
BC052292.1 | - | 7 - 8 | 1475 | 81 | |
BC088354.1 | - | 8 - 9 | 969 | 81 |